VWR® Acrylamide and Bis-Acrylamide
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0254-500MLEA 0 INR
0254-500ML 0172-100G
VWR® Acrylamide and Bis-Acrylamide
Electrophoresis Gels Agarose and Acrylamide
An extensive line of acrylamide and bis-acrylamide pre-weighed powder blends, pre-mixed stock solutions and ready to use solutions for customised polyacrylamide gel electrophoresis of proteins and PAGE of nucleic acids.
Ultra-pure (>99,9%) acrylamide and bis-acrylamide powders provide the flexibility to prepare solutions having concentrations and ratios for all electrophoresis applications.
Liquid blends minimise the handling of neurotoxic acrylamide.
Packaging: Packed in a plastic bottle.