Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent

Supplier: VWR Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:


102923CEA 0 INR InStock
102923C 102927G 102928H 102922B 102926D
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Water
Produced by reversed osmosis and CDI


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098
Order Now
Specification Test Results
Substances reducing KMnO4 Passes test
Conductivity (at filling) Max. 1 µS/cm
TOC (at filling) Max. 400 ppb
pH (25°C) 5.0 - 7.5
Evaporation residue Max. 1 ppm
Ignition residue Max. 1 ppm
Silica Max. 0.1 ppm
Cl (Chloride) Max. 0.05 ppm
F (Fluoride) Max. 0.05 ppm
NH4 (Ammonium) Max. 0.2 ppm
NO3 (Nitrate) Max. 0.01 ppm
PO4 (Phosphate) Max. 0.01 ppm
SO4 (Sulphate) Max. 0.1 ppm
Ag (Silver) Max. 0.0004 ppm
Al (Aluminium) Max. 0.002 ppm
As (Arsenic) Max. 0.002 ppm
Au (Gold) Max. 0.001 ppm
B (Boron) Max. 0.05 ppm
Ba (Barium) Max 0.05 ppm
Be (Beryllium) Max. 0.002 ppm
Bi (Bismuth) Max. 0.001 ppm
Ca (Calcium) Max. 0.05 ppm
Cd (Cadmium) Max. 0.001 ppm
Co (Cobalt) Max. 0.001 ppm
Cr (Chromium) Max. 0.0004 ppm
Cu (Copper) Max. 0.0004 ppm
Fe (Iron) Max. 0.001 ppm
Ga (Gallium) Max. 0.004 ppm
Ge (Germanium) Max. 0.001 ppm
In (Indium) Max. 0.001 ppm
K (Potassium) Max. 0.05 ppm
Li (Lithium) Max. 0.002 ppm
Mg (Magnesium) Max. 0.005 ppm
Mn (Manganese) Max. 0.0004 ppm
Mo (Molybdenum) Max. 0.002 ppm
Na (Sodium) Max. 0.1 ppm
Ni (Nickel) Max. 0.0004 ppm
Pb (Lead) Max. 0.001 ppm
Pd (Palladium) Max. 0.008 ppm
Pt (Platinum) Max. 0.001 ppm
Sb (Antimony) Max. 0.001 ppm
Sn (Tin) Max. 0.001 ppm
Sr (Strontium) Max. 0.0004 ppm
Ti (Titanium) Max. 0.001 ppm
Tl (Thallium) Max. 0.00005 ppm
V (Vanadium) Max. 0.001 ppm
Zn (Zinc) Max. 0.004 ppm
Zr (Zirconium) Max. 0.001 ppm
ISO Standard Water Grade 3 Passes test

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms