Disposable earplugs, Laser Lite®
Supplier: HONEYWELL SAFETY PRODUCTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LL-1
111-2301EA 0 INR
111-2301
Disposable earplugs, Laser Lite®
Earplugs Disposable Earplugs
Laser Lite® earplugs offer optimal comfort for long-term wear.
  • Bright colours for high visibility
  • Self-adjusting foam to fit virtually every user
  • T-shape for easy insertion and wear
SNR: 35 dB
EN 352-2
Certifications: EN 352-2
Caution: * One size fits all.