Cleanroom gloves, latex, sterile, BioClean Maxima™
Supplier: NITRITEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BLLS80
112-3845EA 0 INR
112-3845
Cleanroom gloves, latex, sterile, BioClean Maxima™
Gloves
These cleanroom gloves with beaded cuffs can be used in medical as well as in laboratory environments. Suitable for double-donning.
  • Sterile, powder-free, textured finish
  • Protein level less than 50 µg/g
  • Latex provides exceptional flexibility and comfort
  • Anatomically shaped for enhanced wearer comfort
  • Extended length provides full arm protection
  • Each size carries a unique barcode on the carton (ITF-14), which allows automatic data entry into stock control systems
Suitable for use in Class 10, ISO 4, M2.5 and EU GMP Grade A cleanrooms.
Certifications:More Product Information