Industrial gloves, uvex phynomic C3
Supplier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6008006 6008007 6008008 6008009 6008010 6008011
112-4942EA 0 INR
112-4942 112-4943 112-4944 112-4945 112-4946 112-4947
Industrial gloves, uvex phynomic C3
Gloves
The uvex phynomic is an ultra-light all-round safety glove with good cut protection (Cut 3). The polyamide/elastane/HPPE/glass glove liners feature an aqua-polymer foam coating on palm and fingertips.
  • Foam coating is extremely flexible, provides good grip and leaves no trace on sensitive surfaces
  • Excellent dexterity
  • Very high level of breathability
  • Accelerator-free and suitable for allergy sufferers
Ideal for precision assembly and precision work. Suitable for use in dry and slightly damp areas.
Certifications: EN 388:2016 (4X43B)
Food contact approved
OEKO-Tex® Standard 100