Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® Nitrile Green Nitrile gloves
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
112-5157EA 0 INR InStock
112-5157 112-5155 112-5156 112-5158 112-5154
VWR® Nitrile Green Nitrile gloves
Gloves
These single-use examination and disposable protective gloves are made from nitrile and are developed with sustainability in mind. They are suitable for use in medical applications, food processing and for general lab requirements in non hazardous areas.
  • Free from chlorine, natural rubber latex proteins and rubber accelerators, particularly skin friendly for users with latex allergy
  • Non sterile, powder-free
  • Textured fingertips provide extra grip for wet or dry applications
  • Synthetic inner coating for fast and easy glove donning
  • Comfortable, strong glove
More Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms