Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Cleanroom wipes, sterile, SterileWipe™ LP 10
Supplier: ITW-CONTAMINATION TEXWIPE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
TX3212
115-0312EA 78734.56 INR InStock
115-0312
Cleanroom wipes, sterile, SterileWipe™ LP 10
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
SterileWipe™ LP 10 is a 100% continuous-filament polyester wipe with thermally sealed borders that have been designed to control particle and fiber contamination. The wipe is suitable for cleaning sterile environments such as pharmaceutical aseptic fill areas, biotechnology manufacturing facilities, microbiological laboratories, sterile suites and prep rooms.
  • Extremely low levels of releasable particles and fibres
  • Withstand exposure to common cleaning and disinfecting solutions including bleaches, phenols and quaternary ammonium compounds
  • Sterilised by
More Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms