Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Cleanroom wipes, sterile, Sterile TexTra™
Supplier: ITW-CONTAMINATION TEXWIPE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
TX3220
115-0592EA 93296.68 INR InStock
115-0592
Cleanroom wipes, sterile, Sterile TexTra™
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
Sterile TexTra™ is a single-ply, double-knit, 100% continuousfilament polyester wipe that offers high-capacity sorption in a heavyweight, large surface area wipe. It is an economical alternative to two-ply polyester wipes and ideal for spill control as well as for cleaning pharmaceutical aseptic fill areas, biotechnology manufacturing facilities, and microbiological laboratories.
  • Good abrasion resistance and superior strength
  • Large wipe size and liquid capacity simplify the application and removal of large volumes of cleaning and disinfecting solutions
  • Low
More Product Information
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms