VWR® PUREImage, Cleanroom Papers
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
87004-188 87003-434
115-0149EA 0 INR
115-0149 115-0150 115-0156 115-0163 115-0164 115-0165 115-0166 VWRU87004-188 VWRU87003-434 115-1951 115-1952 115-1953 115-1954 115-1955 115-1956 115-1957 115-1959 115-1960 115-1958 115-1961
VWR® PUREImage, Cleanroom Papers
Paper
Cleanroom paper is designed for use in cleanroom environments, where the prevention of chemical and particle contamination is critical.
  • High tensile strength, and excellent heat resistance, for enhanced copier and printing efficiency
  • PE-binding technology results in very low levels of particles, non volatile residue and ion extractables
  • Unique precision-cutting technology guarantees consistent sizing and cleanliness
  • Paper is web-cleaned on both sides prior to sheeting
Produced in an ISO Class 4 (FED-STD-209E Class 10/M2,5) cleanroom and cleaned andMore Product Information