Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Absorbents - Mats, Rolls and Socks for Oil Spills
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
121-0137EA 0 INR InStock
121-0137 121-0143
VWR®, Absorbents - Mats, Rolls and Socks for Oil Spills
Spill Control Materials
VWR® absorbents and spill-containment products help keep your employees and work environment safe from unexpected spills, leaks, drips or other accidents.
  • Will only absorb oil and petroleum-based liquids
  • Will not absorb water-based liquids
  • Can be used outdoors as they will not absorb water
Colour: White
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms