VWR® Protection Covers
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
129-1210EA 0 INR
129-1210 129-1215 129-1219 129-1218
VWR® Protection Covers
Covers
Manufactured from a specially formulated breathable microporous fabric, these covers are distinguished by their superior barrier protection capabilities.
  • Superior fluid and particulate barrier
  • Lowest level of particle shedding
  • Light chemical splash resistance, and maximum tear strength
  • Tunnelled elastic, along the bottom perimeter, ensures a secure fit
  • Serged seams
VWR® cleanroom protection covers are suitable for GMP/ISO cleanrooms of life science and other critical manufacturing sites. They are customised to fit machines, equipment, and carts in theMore Product Information