Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
HD Super™ process tables
Supplier: METRO
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WT307US
132-2105EA 0 INR InStock
132-2105
HD Super™ process tables
Furniture Tables and Workbenches
These stainless steel laboratory workbenches are available in two versions, with or without a bottom shelf. The versions without a bottom shelf have three transverse struts allowing free footroom without an impeding front bar.
  • Unobstructed flat surface for unimpeded work
  • No sharp edges through specially rounded corners
  • All stationary tables are 864 mm high with stationary posts and levelling feet (feet individually height-adjustable)
  • All mobile tables are 864 mm high with special posts and 4 poly casters (127 mm) - 2 swibel/2 brake
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms