Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

n-Hexane ≥99.0%, SPECTRONORM® for spectroscopy

Supplier: VWR Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger

Filtered through a 0.2 µm filter, packaged under nitrogen.

140096EEA 0 INR InStock
140096E
n-Hexane ≥99.0%, SPECTRONORM® for spectroscopy
n-Hexane
SPECTRONORM solvents have extensive specifications to meet the highly demanding requirements of UV applications.

  • Meets the requirements of "Specific use spectroscopy" according Reag. Ph.Eur.


Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: –94,3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
REACH: 01-2119480412-44
Merck Index: 13,04712
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c939cebcd37af6287105861069a46c7635aa4af8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay (GC) ≥ 99.0 %
Acidity ≤ 0.0002 meq/g
Alkalinity ≤ 0.0002 meq/g
Colouration ≤ 10 APHA
Evaporation residue ≤ 2 ppm
Water ≤ 50 ppm
Absorbance (195 nm) ≤ 1.00
Absorbance (210 nm) ≤ 0.22
Absorbance (217 nm) ≤ 0.10
Absorbance (225 nm) ≤ 0.05
Absorbance (from 245 nm) ≤ 0.01
Transmittance (195 nm) ≥ 10 %
Transmittance (210 nm) ≥ 60 %
Transmittance (217 nm) ≥ 80 %
Transmittance (225 nm) ≥ 90 %
Transmittance (from 245 nm) ≥ 98 %
Fluorescence (as quinine) (254 nm) ≤ 1.0 ppb
Fluorescence (as quinine) (365 nm) ≤ 1.0 ppb

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms