Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® Critical Swabs
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
89031-270
149-0330EA 0 INR InStock
149-0330 149-0331 149-0332 149-0340 149-0341 149-0342 VWRU89031-270
VWR® Critical Swabs
Swabs Swabs for Sampling
Critical swabs designed for cleaning disc drives, optics, PC board assemblies and other sensitive components.
  • Highly absorbent
  • Low linting formulation reduces contamination
  • Double-headed items in a white, anti-static leaf pack
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms