Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Chromatography Tubing, PTFE
Supplier: HICHROM
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
08-PTFE-62-3MEA 0 INR InStock
08-PTFE-62-3M 16-PTFE-07-3M 16-PTFE-20-3M 16-PTFE-30-3M 08-PTFE-62-10M 16-PTFE-07-10M 16-PTFE-20-10M 16-PTFE-30-10M 08-PTFE-62-5M 16-PTFE-07-5M 16-PTFE-20-5M 16-PTFE-30-5M
Chromatography Tubing, PTFE
Tubing Chromatography Tubing
PTFE
  • Flexible and economical
  • Excellent solvent resistance
Inert to all chemicals used in HPLC but suitable for low-pressure applications only. The most common tubing used to connect the solvent reservoir to the pump.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms