Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® Deioniser with Interchangeable Cartridges
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
171-2708EA 0 INR InStock
171-2708
VWR® Deioniser with Interchangeable Cartridges
Water Purification Systems Deionisers
This demineraliser connects to a mains water tap and instantly delivers pure water with high resistivity. The exchange capacity of a cartridge is 300 l of water for a town mains water supply at 25° hydrometric.
  • Easy to use
  • Economic solution
  • Easy cartridge change (change in colour when the resins are spent)
The resistivity of the water produced is always greatly superior to 1 MΏ/cm: it normally rises to 10 MΏ/cm after drawing off the first 30 litres. The normal flow rate is 20 to 40 L/h, with a maximum of 80 L/h.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms