Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Diaphragm valves, VM series
Supplier: VACUUBRAND
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
20664010 20664011
183-1151EA 0 INR InStock
183-1151 183-1152
Diaphragm valves, VM series
Valves Vacuum Valves
VM series valves are hand operated diaphragm valves. They are used as isolation valves, air-admittance valves or manual-control valves.
  • Materials with high chemical resistance
  • High conductance, low leakage rate
  • Flow control possible
  • Gas-tight against atmosphere in both directions
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms