Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Flasks, Flat Bottom, Narrow or Wide Neck
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
214-1187EA 0 INR InStock
214-1187 214-1188 214-1189 214-1190 214-1191 214-1192 214-1193 214-1194 214-1195 214-1196
VWR®, Flasks, Flat Bottom, Narrow or Wide Neck
Flasks Flat Bottom Flasks
Borosilicate glass 3,3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Marked with capacity and a white labelling area.
ISO 1773 (narrow mouth)
DIN 12347 (wide mouth)
Certifications: ISO 1773 (narrow neck)
DIN 12347 (wide neck)
Order Now
ORDER
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms