Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Chemical spoon, double-ended
Supplier: RSG
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
023.120
231-1221EA 0 INR InStock
231-1221
Chemical spoon, double-ended
Spoons
Stainless steel 18/10, anti-magnetic. Multipurpose spoon for a wide range of uses. With cylindrical handle.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms