32040.298EA
9181.73
INR
InStock
32040.298
32096.291
32031.297
32032.291
32033.294
32033.374
32035.291
32036.294
32038.291
32039.261
32039.374
32040.185
32040.378
32041.292
32042.295
32044.268
32044.292
32044.372
32045.262
32045.295
32045.375
32046.265
32046.298
32046.378
32095.184
32095.264
32095.366
32095.377
32096.187
32096.267
32096.360
32096.371
32097.261
98192.290
98213.260
32039.294
32095.297
83601.260
83601.290
83610.600
32032.260
98192.260
83610.610
32097.290
32040.260
32096.400
32044.290
32045.290
32045.412
32044.418
85680.290
85680.295
85680.260
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Buffers
pH Calibration Buffers
Solutions prepared from AnalaR® NORMAPUR® grade analytical reagents.
- Reference temperature 20 °C
- True analytical value on the label to an accuracy of ±0,02 pH units (pH 1,00 to 11,00); ±0,05 for pH 12,00 buffer
- Traceable to SRM from NIST
- Batch number and packing date for traceability
- Certificate of Analysis available at website
Manufactured from analytical grade products presenting high accuracy: 0,02 pH unit from pH 1,00 to 11,00 and 0,05 pH unit for pH 12,0.
Certifications: Certifications: Products with accreditation to ISO 17034 in the table below are produced and certified in accordance with ISO 17034 accredited by BELAC under the certification number 542-RM. All information regarding the use of these CRMs can be found on the Certificate of Analysis.
Ordering information: If significantly large volumes are required please contact your local Avantor sales office for a customised offer.
Packaging: Available in different pack sizes: 100 ml, 500 ml, 1 L, 5 L plastic bottles and bag-in-box (5 and 10 L).