VWR® INCU-Line®, Shaking Incubators
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
444-0732EA 682147.62 INR
444-0732 444-0733
VWR® INCU-Line®, Shaking Incubators
Shakers Shaking Incubators
Shaking incubators with a small footprint. These compact units feature a stackable design and expanded temperature range which makes the ILS 4 and ILS 6 ideal for molecular biology applications and general use. The integral orbital shaker means they are highly versatile. The SmartChek™ temperature control system (exclusive to the Avantor Collection) guarantees precise temperature control. Mechanical convection maintains a stable temperature environment and provides quick recovery after opening the door. A safety thermostat is located on the back of the unit. The insulated door has a large,More Product Information