Microplate shakers, Titramax range
Supplier: Heidolph Instruments GmbH & Co.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
544-11300-06 544-11200-13 544-12200-06 544-12209-06 544-11200-02 544-12200-02 544-12209-02
444-0195EA 0 INR
444-0195 444-0194 444-0196 444-0190 444-1483 444-1485 444-1486
Microplate shakers, Titramax range
Shakers Microplate Shakers
For vigorous mixing of microtitre plates. Vibration orbit of 1,5 mm guarantees no spills even with full vessels, 3 mm orbit gives best mixing results even with the smallest quantities. The shakers are designed for continuous operation and feature over-temperature protection, and an insulated platform with virtually no heat-up.
  • Modular system, Titramax 1000 can be combined with the Incubator 1000 and a flat hood
  • Timer function with audible signal allows a process time up to 120 minutes
  • Electronic speed control
  • Ideal for use under controlled conditions, can
More Product Information