VWR® Advanced 700, Magnetic Hotplate Stirrer
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
444-2862EA 0 INR
444-2862 444-2866 444-2868 444-2870
VWR® Advanced 700, Magnetic Hotplate Stirrer
Stirrers Magnetic Hotplate Stirrers
VWR® 700 Series Advanced hotplate stirrers deliver accurate and repeatable results. Low profile, space saving design takes up less space in the laboratory. Spill-resistant housing channels fluids away from internal components. Front panel features easy to use controls, which allow users to dial-in adjustments. Integral support rod holder with locking knob accepts the optional support rod and clamp kit. Microprocessor control regulates heating and stirring functions. Samples are heated to temperature quickly and accurately. Speed is precisely controlled, giving constant stirring throughoutMore Product Information