Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® Signature 800, Hotplates
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
444-2871EA 0 INR InStock
444-2871 444-2873 444-2875 444-2877
VWR® Signature 800, Hotplates
Heaters Hotplates
VWR® Signature hotplates are designed for applications that require exceptional accuracy, stability and repeatability.
  • Separate LED displays for temperature and time
  • Includes external temperature probe and support rod kit
  • Range of safety features protects operator and sample
  • Ceramic or aluminium top plates with hot warning
Microprocessor control regulates heating function. It also includes an external probe option that gives ±1% temperature stability. A spill resistant housing channels fluids away from internal components.
Safety features includeMore Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms