Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Vortex mixers, Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SI-0256 SI-0156 SI-T256 SI-0276 SI-0176 SI-0266 SI-0166
444-5900EA 65319.73 INR InStock
444-5900 444-9173 444-9174 444-9182 444-9183 444-0486 444-0485
Vortex mixers, Vortex-Genie®
Shakers Vortex Shakers
Rugged and reliable vortex mixers with solid metal casing and rubber feet that prevent them 'walking'. Vortex-Genie® 1 features touch-on operation and fixed speed, it is ideal for aggressive vortexing of difficult samples.
  • Wide range of attachments available
  • Vortex-Genie® 2T timer function; touch-on 1 to 60 seconds, hands-free 1 to 60 minutes or continuous
  • Can be used in cold room or incubator
Vortex-Genie® 2 has variable speed for precise mixing from gentle to vigorous, it offers hands-free or touch-on control. Mixing action prevents spilling, evenMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms