Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Stainless Steel Sand Baths
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
460-3227EA 28735.56 INR InStock
460-3227 460-3228 460-3229 460-3230 460-3231 460-3232 460-3233
VWR®, Stainless Steel Sand Baths
Heaters Sandbaths
Ideal for irregular-shaped vessels. Stainless steel construction for superior corrosion resistance. Designed to hold sand, stainless steel shot or non volatile liquids.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms