Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Bath fluids
Supplier: JULABO GmbH
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
8940115 8940101 8940103 8940104 8940105 8940107 8940109 8940102 8940133 8940132 8940141
461-0226EA 0 INR InStock
461-0226 461-0110 461-0120 461-0131 461-0130 461-0140 461-0225 461-0121 461-0478 461-0477 461-0878
Bath fluids
Circulator Bath Fluids
Bath fluids have been carefully selected and proven in long-term service. They are suitable for use with heating and refrigerated circulators whenever water cannot be employed as bath fluid. The choice of bath fluid is critical for best results in temperature control and operation of the circulator.
  • Low toxicity and almost odourless
  • High stability and good thermal conductivity
  • Low viscosity and low corrosiveness
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms