Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Desiccator bases, DURAN®
Supplier: DWK LIFE SCIENCES
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
247704604 247705703 247706108 247706605 247706905 247714605 247715704 247716109 247716606 247716906 247725705 247726101 247726607 247726907
467-1110EA 0 INR InStock
467-1110 467-1115 467-1120 467-1125 467-1130 467-1210 467-1215 467-1220 467-1225 467-1230 467-1315 467-1320 467-1325 467-1330
Desiccator bases, DURAN®
Desiccators Desiccator Bases
DURAN®, clear borosilicate glass 3.3. Ideal for drying and drying processes, storage under vacuum and protection from the effects of air and moisture. Applications include cultivation of organisms, material research and corrosion testing, cleaning equipment for analysis of trace elements, gas and liquid reactions and they can be used as a mini cleanroom for chip technology.
  • Choice of various desiccator components to suit different applications
  • Increased safety due to DURAN® quality borosilicate glass
  • All models have a flat flange, except 467-0139 which
More Product Information
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms