Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Laboratory freezers, MediLine, LGPv series
Supplier: Liebherr
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LGPV1420 LGPV1430 LGPV6530 LGPV8420 LGPV8430
472-2389EA 0 INR InStock
472-2389 471-0661 471-0662 472-0118 471-1017
Laboratory freezers, MediLine, LGPv series
Freezers Laboratory / General Purpose Freezers
These are heavy duty MediLine models with forced air cooling. The controller features door and temperature alarms which can be customised to suit the end user. Controller can be locked. The refrigerant used is hydrocarbon. Units have 83 to 60 mm insulation and 2 castors with brakes.
  • Back-up battery for 7,5 days data logging and 72 hours battery back up for controller in case of mains failure
  • RS485 interface connection, volt-free contacts, pre-drilled access port and optional infrared key for data download
  • Forced air ventilation with hot gas defrost - for
More Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms