VWR®, Test Sieves, 400×65 mm
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
510-3478EA 0 INR
510-3478 510-3479 510-3480 510-3481 510-3482 510-3483 510-3484 510-3485 510-3486 510-3487 510-3488 510-3489 510-3490 510-3491 510-3492 510-3493 510-3494 510-3495 510-3496 510-3497 510-3498 510-3499 510-3500 510-3501 510-3502 510-3503 510-3504 510-3505 510-3506 510-3507 510-3508 510-3509 510-3510 510-3511 510-3512 510-3513 510-3514 510-3515 510-3516 510-3517 510-3518 510-3519 510-3520 510-3521 510-3522 510-3523 510-3524 510-3525 510-3526 510-3527 510-3528 510-3529 510-3530 510-3531 510-3532 510-3533 510-3534 510-3535 510-3536 510-3537 510-3538 510-3539 510-3540 510-3541 510-3542 510-3543 510-3544 510-3545 510-3546 510-3547 510-3548 510-3549 510-3550 510-3551 510-3552 510-3553 510-3554 510-3555 510-3556 510-3557 510-3558 510-3559 510-3560 510-3561 510-3562 510-3563 510-3564 510-3565 510-3566 510-3567 510-3568 510-3569
VWR®, Test Sieves, 400×65 mm
Sieves Test Sieves
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.