Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Tangential flow systems, Vivaflow® 50 and 200
Supplier: Sartorius
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VF05P4 VF20P0 VF20P1 VF05P9
512-4065EA 0 INR InStock
512-4065 512-4069 511-0876 515-0142
Tangential flow systems, Vivaflow® 50 and 200
Filters Tangential Flow Cassettes
Vivaflow® 50 and 200 devices are ready to use crossflow cassettes for filtration and concentration of samples from 100 to 3000 ml or 500 to 5000 ml. The membrane surface per device is 50 cm² or 200 cm² respectively. The devices come with all the necessary accessories for running with a laboratory pump and a size 16 pump head. For speeding up the concentration and increasing the volume throughput, up to six Vivaflow® 50 cassettes can be connected together or two Vivaflow® 200 can be connected together. Optimally suited for applications like cell culture supernatant concentration, virusMore Product Information
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms