Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Personal Micro-Centrifuge
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
521-2778EA 9939.5 INR InStock
521-2778
VWR®, Personal Micro-Centrifuge
Centrifuges Microcentrifuges
Made from the highest quality of raw materials and components, this micro-centrifuge will fit seamlessly into your work life and deliver exceptional performance. Designed to offer optimum performance and years of durability, the unit has been especially tailored to provide enhanced airflow to prevent the rise of temperature inside the chamber.
  • Electric brake for stopping the device immediately
  • Magnetic design for tool-free rotor removal
  • Carbon dust insulation and safe drainage provision
  • Quick run function to save time
  • Speed accuracy of ±200 min⁻¹
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms