Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Digital disc rotators
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
521-2779EA 38496.25 INR InStock
521-2779 521-2780 521-2781
VWR®, Digital disc rotators
Shakers Rotating/Rocking Shakers
The Digital disc rotator is the result of extensive research and user experience feedback. Consisting of a maintenance free brushless DC motor, the rotator offers powerful performance without any hassel. The features such as intermittent mode, user programmable setup and its pulse mode offer thorough mixing without much effort.
  • Microprocessor controlled disc rotator for intelligent performance with last run memory feature
  • Variable angle setting to adjust the rotator angle from 0 to 90°
  • User programmable mode to set 9 user-defined programs for greater
More Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms