Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Gastight® 1700 series syringes
Supplier: HAMILTON BONADUZ
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
80000 80030 80075 80201 80230 80901 80930 81020 81030 81039 81101 81120 81130 81217 81220 81243 CAL81020 80016 CAL81220
549-1124EA 0 INR InStock
549-1124 549-1126 549-0220 549-1131 549-1132 549-1170 549-1171 549-0365 549-1176 549-0366 549-1180 549-0367 549-1181 549-1186 613-1425 549-0368 548-1974 548-2285 548-1975
Gastight® 1700 series syringes
Syringes Syringes for Chromatography
These syringes are the Gastight® version of the original 700 series syringes and are ideal for dispensing volumes from 1 µl up to 500 µl.
  • Fluid path includes stainless steel, borosilicate glass and PTFE
  • Gas sterilisable
Gastight® syringes are ideal for dispensing both liquids and gases and have a precision machined PTFE plunger tip which creates a leak free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that aMore Product Information
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms