Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Modified syringes Microliter™7000 series
Supplier: HAMILTON BONADUZ
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
86250 86259 88400 80135
549-1226EA 0 INR InStock
549-1226 549-1227 549-1237 549-1128
Modified syringes Microliter™7000 series
Syringes Syringes for Chromatography
Suitable for applications such as GC, TLC and micro pipetting. Manufactured according to the principle of dead volume by positive pressure. A tungsten piston is introduced into the syringe as far as the tip of the needle. Needle and piston are calibrated against each other and must be changed at the same time in the event of replacement.
  • Volumetric dosage takes place in the needle
  • Precision ±1% of the nominal volume
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms