Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® Classic LP / LPC / LPW, Precision Balances with LCD
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
611-2610EA 67802.28 INR InStock
611-2610 611-2301 611-2300 611-2298 611-2302 611-2296 611-2289 611-2308 611-2279 611-2320 611-2272 611-2278 611-2275 611-2274 611-2321 611-2319 611-2629 611-3269 611-3270 611-3271 611-3272 611-3277 611-3283 611-3296 611-3298 611-3327 611-3379 611-3346 611-3347 611-3348 611-3354 611-3355 611-3359 611-3360 611-3361 611-3367 611-3371 611-3373 611-3374 611-3375 611-3383 611-3395 611-3397 611-3398 611-3458 611-3472 611-2600 611-3816 611-3823 611-3825 611-4316 611-4317 611-4330 611-5616 611-5617
VWR® Classic LP / LPC / LPW, Precision Balances with LCD
Balances and Scales
Rugged and economical balances, that are ideal for educational establishments and laboratory use. A series that ranges from simple weighing to operation with automated internal calibration. All Classic models have a simple backlit LCD with adjustable contrast and ergonomic keypad and a stainless steel top pan.
  • Selectable filter levels and user can select the measuring unit that the balance will use to display the weight
  • RS232 data interface
  • LPW models have a three door draft shield
Functions include suspended weighing, piece counting, percentageMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms