Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Semi-Micro Balances with Ioniser, SM and SMG Series
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
611-3801EA 232218.84 INR InStock
611-3801 611-3803 611-3804 611-3806 611-3807 611-3809 611-3810 611-3800 611-4284 611-4294 611-4314 611-4315
VWR®, Semi-Micro Balances with Ioniser, SM and SMG Series
Balances and Scales
Semi-micro balances with resolution of 0,01 mg and a built-in ioniser, which ensures accurate sample preparation. Static charges accumulate in different samples that are routinely weighed in the laboratory. These static charges can build up and alter the weighing value. By blowing ions, the built-in ioniser neutralises static electricity and within seconds the sample is ready to be weighed. The balance calibration system automatically calibrates the balance when there is a temperature change or at a set period of time (factory settings), this ensures accurate measurements even if the userMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms