Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Bottle top burettes
Supplier: VITLAB
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1620506
613-2167EA 0 INR InStock
613-2167
Bottle top burettes
Bottle Top Dispensers
The continuous bottle-top burette enables continuous titration, which leads to rapid, convenient, and accurate results.
  • Long operating life from two easily-replaced micro-batteries 1,5 V
  • Large, easy-to-read digits
  • Simple, media-specific readjustment via keypad
  • Continuous and precise regulation of titration speed with a large practical thumbwheel
  • Reduced titrant waste with innovative recirculation valve
  • Adaptable to bottles of different sizes
  • Telescopic discharge tube which can be adjusted in both height and length
  • Telescopic, extractable intake tube adapts
More Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms