Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Calibrated electronic verification thermometers, frio temp
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
620-0904EA 10207.68 INR InStock
620-0904 620-2148 620-2344
Calibrated electronic verification thermometers, frio temp
Thermometers Digital Thermometers
Digital thermometers verify sample temperatures in freezers, refrigerators, incubators and ovens. Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample.
  • Individually serialised
Plastic bottles cannot be used in ovens.
Certifications: ISO 17025, ISO/IEC 17025:2005, traceable to NIST.
Delivery information: Includes multi-language instructions (English, French, German, Italian, Portuguese and Spanish), AAA or LR44 (620-2148) batteries.
Caution: * Internal
** External
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms