VWR® Safety-Blue™, Thermo-Hydrometers
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
620-0924EA 0 INR
620-0924 620-0925 620-0926 620-0928 620-0929 620-0932
VWR® Safety-Blue™, Thermo-Hydrometers
Hydrometers
These safety sugar scale combined form thermo-hydrometers can determine the specific gravity, Brix and alcohol by volume.
  • Lead free ballast and glass with mercury-free blue spirit organic liquid fill
  • Individually serialised, safe and easy to clean up in the event of a breakage
  • Temperature standardisation of 20 °C with thermometer
Designed to measure sucrose in the food and beverage and other industries.
Certifications: ASTM
Delivery information: Includes a multi-language statement of accuracy indicating traceability to NIST.
Caution: CombinedMore Product Information