Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
TRACEABLE MEMORY-LOC LN2 1 * 1 items
Supplier: VWR Collection
Digital thermometer, VWR® Traceable®, With 1 probe, Insertion or penetration probe, Measuring channels: 1, Probe: 1 stainless steel, Range: −200...+105 °C, Accuracy: ±0,25 °C, Resolution: 0,01 °C, Weight: 141,74 g, Supplied with calibration certificate
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Quantity
TRACEABLE MEMORY-LOC LN2 1 * 1 items 620-2635 1 * 1 items
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Print These Terms