VWR® Traceable® Digital Heat Thermometer
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
620-2809EA 6209.28 INR
620-2809
VWR® Traceable® Digital Heat Thermometer
Thermometers Digital Thermometers
Simultaneously displays temperature, timer, and current alarms.
  • Tri-display temperature, alarm, timer
  • Programmable high/low alarm
  • ºC/ºF exchange
  • 12/24 hour countdown timer
  • Timer setting memory
  • Timer elapse time display
  • Stainless steel probe
  • Magnetic attachment
  • Low battery indicator
Perfect for use in ovens and incubators.
Note: New product design for 620-1912.
Certifications: Individually calibrated ISO 17025 certified. Individually certified—traceable to NIST.