Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® VisiScope® 360, Stereo Zoom Microscopes
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
630-3090EA 73132.5 INR InStock
630-3090 630-3091 630-3085 630-3084 630-3087 630-3089 630-3086 630-3088
VWR® VisiScope® 360, Stereo Zoom Microscopes
Microscopes
Choose from a range of binocular and trinocular stereo zoom microscope heads for either basic or advanced requirements.
  • Parfocal achromatic zoom
  • Greenough optical system
  • Available with or without illumination
  • Stable, slim modern stand or long overhanging stand for observation of particularly large specimens
  • Extended zoom range
Coarse focusing system (76 mm head holder, 32 mm pillar) with adjustable tension for standard stereo microscope requirements. Binocular head or trinocular head (split ratio: 50/50) feature 360° rotating on all stands and 45°More Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms