VWR® VisiLight® LED8, High Power Cold Light Source
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
631-1258EA 136545.36 INR
631-1258
VWR® VisiLight® LED8, High Power Cold Light Source
Lights Cold Light Sources
This cold light source with compact and stable aluminium housing and temperature fan control provides an optimal and homogenous illumination of the light guide. It is therefore ideal for high speed recordings in a variety of applications, such as quality assurance, industrial endoscopy, machine vision, and cleanroom applications. The brightness exceeds by far the brightness of a conventional 250 W halogen light source. Expected LED lifespan of 50000 hours.
  • 1600 lm at the end of the light guide
  • Temperature fan control for minimal noise disruption
  • Analogue
More Product Information