Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Bench Top Refractometers with Peltier Temperature Control, PLR1/PLR2
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
635-0844EA 1087800 INR InStock
635-0844 635-0845
VWR®, Bench Top Refractometers with Peltier Temperature Control, PLR1/PLR2
Refractometers Benchtop Refractometers
Simple to use, precise bench top refractometers for use in laboratory or production environments.
  • Wide measuring range
  • Brix, RI, user scales
  • Peltier temperature control
  • 7" touch screen
  • Multilingual display
  • Intuitive menu structure
  • Store up to 4000 readings
Two distinct high precision bench top refractometers that offer users a choice based on measurement range and application type. Both models feature refractive index (nD) and sugar (Brix) scales and it is possible to load up to 25 commonly used scales from the instrument’s library.
The PLR1 isMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms