J.T.Baker® BAKERBOND®, SPE 96-Well Plates
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7020-P10EA 0 INR
7020-P10 7090-P10 7091-P10 7089-P10 7087-P10 7086-P10 7211-P10 7225-P10 7466-P10 7020-P30 7090-P30 7091-P30 7089-P30 7087-P30 7086-P30 7211-P30 7225-P30 7466-P30 7020-P60 7090-P60 7091-P60 7089-P60 7087-P60 7086-P60 7211-P60 7225-P60 7466-P60
J.T.Baker® BAKERBOND®, SPE 96-Well Plates
Microplates Solid Phase Extraction Plates (SPE)
Solid Phase Extraction (SPE) is commonly used as a highly selective sample clean-up or sample enrichment method prior to subsequent analyses. J.T.Baker® BAKERBOND® SPE 96 well plates are well suited for high throughput and automated SPE methods. With complementary high capacity sorbents to existing spe extraction columns in the J.T.Baker® BAKERBOND® range, higher throughput methodologies can now be simply developed and automated.
  • 96 well format for multiple sample preparation
  • Consistent sorbent weight to reduce variability
  • Different chemistries available
More Product Information