Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
J.T.Baker® BAKERBOND® ABx™, Ion Exchange Chromatography Media
Supplier: AVANTOR PERFORMANCE MATERIALS
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7157-02EA 99211.28 INR InStock
7157-02 7269-02 7269-05 7269-07 7269-01 7369-02 7269-03
J.T.Baker® BAKERBOND® ABx™, Ion Exchange Chromatography Media
Chromatography Sorbents HPLC Sorbents
Prep-scale packing is intended for use at low pressure with a peristaltic pump.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms