Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Filter Tips, Low Retention
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
732-1488EA 8338 INR InStock
732-1488
VWR®, Filter Tips, Low Retention
Pipette Tips
Low retention tips made of transparent PP resins offering unsurpassed fluid retention performance and proven aerosol blocking with endotoxin-free, 10 μm porous, hydrophobic filter. Ideal for PCR and DNA manipulation.
  • Sustainable design based on virgin resins
  • With PE filter, sterile
  • Up to 10 times less fluid retention than standard tips
  • Sample can be completely recovered without cutting into tip
  • Lot-certified purity (free from protease, RNase/DNase, human DNA and endotoxin, non pyrogenic)
  • Tips and racks are autoclavable to 121 °C for a maximum of 1
More Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms