Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Thermal Cyclers
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
732-2548EA 1063374.48 INR InStock
732-2548 732-2549 732-2550 732-2551 732-2552 732-2554 732-2555 732-2914 732-2915
VWR®, Thermal Cyclers
PCR Cyclers
The VWR® thermal cycler family combines high quality engineering with a comprehensive range of block formats. The UNO cycler is designed around a powerful, yet easy to use, software interface, and uses the same platform for both 96-well and 384-well formats. The Doppio, with two independent high speed 48-well blocks in one system, offers the optimum solution for maximum flexibility within a minimum footprint. Because of the special design of the heated lid, the height adjusts automatically to the different tube sizes.
  • Outstanding performance: Thermal plate with impressive thermal
More Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms