Peqlab peqGOLD SafeGuard™, Filter Tips
Supplier: VWR Peqlab
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
732-2969EA 0 INR
732-2969
Peqlab peqGOLD SafeGuard™, Filter Tips
Pipette Tips
SafeGuard™ filter tips use a micropore technology which actively prevents aerosols reaching and contaminating the pipette. The filter itself is manufactured from PE, which has extremely hydrophobic properties and therefore does not require additional chemicals used in some filter tips which carry the risk of sample contamination. Furthermore, micropore materials allow for full sample recovery in the event of over pipetting.
  • Certified free of RNases, DNases, human DNA, pyrogens and PCR inhibitors
  • Filter reliably prevents the exchange of aerosols between the
More Product Information